Learn More
Sperian™ Honeywell™ Premium Disposable Respirators
Product Code. 11816960
Change view
Click to view available options
Filter Type:
FFP1
FFP2
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type With Exhalation Valve unitSize
11816960 FFP1 No Pack of 20
11806970 FFP2 Yes Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11816960 Supplier Sperian™ Supplier No. 1005580
  • ONLINE EXCLUSIVE PRICE
    44.10 €
    28.90 € / Pack of 20
    Save 15.20 €
    34% Off
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
Highly comfortable and ergonomically designed single-use respirators with no restriction with regard to use. Sperian™ Honeywell™ Premium Disposable Respirators leverage the Willtech™ soft seal technology for an improved fit and exceptional comfort.
  • Rigid shell so shape is maintained even in moist and heavy loading environments
  • Hyopallergenic flange seal with absorption properties
  • Preformed and colored nose bridge to save time
  • Large internal volume to easily communicate and breathe
  • Twin elastic straps, ultra flexible and resistant, stapled outside of the filtering area
  • No latex, no silicone, no PVC
  • Protection and wearer acceptance guaranteed during periods of extended use
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 149:2001A1:2009, ISO 9001 / 2000, EC
Disposable Single-use
Filter Type FFP1
Product Type Respiratory Protection
Size Medium
With Exhalation Valve No
For Use With (Application) Textile, construction, iron and steel industries, etc
Latex Free Yes
Model 5110 M/L Respirator
Quantity 20/Pk.
Resistant To Dusts, Mists, Fumes
Shape Molded Cup