missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, MedChemExpress

Product Code. 16498946
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498946 100 mg 100mg
16484717 50 mg 50mg
16461476 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498946 Supplier Medchem Express Supplier No. HYN0003
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Honokiol is a bioactive, biphenolic phytochemical that possesses potent antioxidative, anti-inflammatory, antiangiogenic, and anticancer activities by targeting a variety of signaling molecules. It inhibits the activation of Akt. Honokiol can readily cross the blood brain barrier.

Honokiol is a bioactive, biphenolic phytochemical that possesses potent antioxidative, anti-inflammatory, antiangiogenic, and anticancer activities by targeting a variety of signaling molecules. It inhibits the activation of Akt. Honokiol can readily cross the blood brain barrier.

Specifications

Chemical Name or Material Honokiol
CAS 35354-74-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18O2
Quantity 100 mg
Synonym NSC 293100
Solubility Information DMSO : ≥ 50 mg/mL (187.74 mM)
SMILES OC1=C(CC=C)C=C(C2=CC(CC=C)=CC=C2O)C=C1
Molecular Weight (g/mol) 266.33
Formula Weight 266.33
Percent Purity 98.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.