missing translation for 'onlineSavingsMsg'
Learn More

HOXA7 Mouse anti-Human, CoraLite 488, Clone: 2A7H6, Proteintech

Product Code. p-7227610 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17791299 20 μL 20µL
17639876 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17791299 Supplier Proteintech Supplier No. CL4886711220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HOXA7 Monoclonal antibody apecifically detetects HOXA7 antigen in Human Samples. It is validated for Immunofluorescence.

Specifications

Antigen HOXA7
Applications Immunofluorescence
Classification Monoclonal
Clone 2A7H6
Concentration 1000 ug/mL
Conjugate CoraLite 488
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC148692
Gene Symbols HOXA7
Host Species Mouse
Immunogen HOXA7 fusion protein Ag19025
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3204
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less
compliance-icons
Product Identifier
  • 20UL CoraLite488-conjugated HOXA7 Mouse McAb
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1A
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.