Learn More
Protein kinase Y linked Antibody (OTI3E10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44632
Product Code. 30306645
  • 735.00 € / 100µg
Description
Protein kinase Y linked Monoclonal antibody specifically detects Protein kinase Y linked in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein kinase Y linked
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
OTI3E10
Western Blot 1:500, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence 1:100
EC 2.7.11, EC 2.7.11.1, protein kinase, Y-linked
Full-length protein expressed in 293T cell transfected with human PRKY expression vector
100 μg
Protein Kinase, Wnt Signaling Pathway
5616
Human
Antibody
IgG2a