missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD9 Antibody (OTI4H10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | HOXD9 |
|---|---|
| Clone | OTI4H10 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306226
|
Novus Biologicals
NBP3-44998-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306490
|
Novus Biologicals
NBP3-44998-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXD9 Monoclonal antibody specifically detects HOXD9 in Human samples. It is validated for Western BlotSpecifications
| HOXD9 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 3235 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI4H10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| homeo box D9, homeobox D9, Homeobox protein Hox-4C, Homeobox protein Hox-5.2, homeobox protein Hox-D9, HOX4, Hox-4.3, mouse, homolog of, HOX4CHox-4.3, Hox-5.2 | |
| Human recombinant protein fragment corresponding to amino acids 109-352 of human HOXD9 (NP_055028) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title