Learn More
Invitrogen™ HOXD9 Monoclonal Antibody (OTI1H3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527194
Product Code. 15956784
  • 570.00 € / 100µL
Description
HOXD9 Monoclonal Antibody for Western Blot, IHC (P)
HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HOXD9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P28356
HOXD9
Human recombinant protein fragment corresponding to amino acids 109-352 of HOXD9 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1H3
Unconjugated
HOXD9
homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9
Mouse
Affinity Chromatography
RUO
3235
-20°C, Avoid Freeze/Thaw Cycles