missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMMPRIN/CD147 Antibody (OX-47), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64848DL594
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| DyLight 594 | |
| 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| T blasts from a mixed lymphocyte reaction. | |
| 0.1 mL | |
| Cancer, Signal Transduction, Vision | |
| 682 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OX-47 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction