Learn More
HR Mouse anti-Human, Clone: OTI1D9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806882
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Drosophila genes can be categorized based on the type of protein for which they encode and are represented by six major classifications, which include intracellular signaling proteins, transmembrane proteins, RNA binding proteins, secreted factors, transcription regulators (basic helix-loop-helix, homeodomain containing, zinc finger containing and chromatin associated) or other functional proteins. Among these numerous proteins, Hairless (CG5460, CT36411, CT17282) is a transcriptional co-repressor that contains a ""Paired box"" domain, a KX-repeat domain, a NEDL-repeat domain and a serine-rich domain. Hairless is an antagonist of the Notch signaling pathway in Drosophila.Specifications
| HR | |
| Monoclonal | |
| Unconjugated | |
| HR | |
| ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55806 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1D9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O43593 | |
| HR | |
| Human recombit protein fragment corresponding to amino acids 946-1189 of human HR produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.