missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSD17B11 Polyclonal Antibody

Description
Peptide sequence: TGEIVLITGA GHGIGRLTAY EFAKLKSKLV LWDINKHGLE ETAAKCKGLG Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 92%.
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
Specifications
Specifications
| Antigen | HSD17B11 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | HSD17B11 |
| Gene Accession No. | Q8NBQ5, Q9EQ06, Q6AYS8 |
| Gene Alias | 17-beta-HSD 11; 17-beta-HSD XI; 17betaHSD11; 17-BETA-HSD11; 17beta-HSD11; 17betaHSDXI; 17-BETA-HSDXI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; 17bHSD11; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; dehydrogenase/reductase (SDR family) member 8; dehydrogenase/reductase SDR family member 8; Dhrs8; estradiol 17-beta-dehydrogenase 11; estradiol 17-beta-dehydrogenase 11-like protein; Hsd17b11; hydroxysteroid (17-beta) dehydrogenase 11; hydroxysteroid 17-beta dehydrogenase 11; MGC137132 protein; PAN1B; PSEC0029; Retinal short-chain dehydrogenase/reductase 2; retinal short-chain dehydrogenase/reductase SDR2; retSDR2; SDR16C2; SDR2; short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2; UNQ207/PRO233 |
| Gene Symbols | HSD17B11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?