missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B13 Antibody (HSD17B13/13102), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44607-100ug
This item is not returnable.
View return policy
Description
HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for nan
Specifications
| HSD17B13 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| HSD17B13/13102 | |
| Unconjugated | |
| HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014 | |
| Recombinant fragment (around aa1-200) of human HSD17B13 protein (Uniprot: Q7Z5P4). Exact sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 345275 | |
| Store at 4⪚C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction