missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B13 Antibody (HSD17B13/13108), Novus Biologicals™
Mouse Monoclonal Antibody
256.00 € - 484.00 €
Specifications
| Antigen | HSD17B13 |
|---|---|
| Clone | HSD17B13/13108 |
| Concentration | 0.2 mg/ml |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
30304441
|
Novus Biologicals
NBP3-44610-20ug |
20 μg |
256.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30295647
|
Novus Biologicals
NBP3-44610-100ug |
100 μg |
484.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for nanTekniske data
| HSD17B13 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014 | |
| Recombinant fragment (around aa1-200) of human HSD17B13 protein (Uniprot: Q7Z5P4). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| HSD17B13/13108 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 345275 | |
| IgG2b κ | |
| Protein A purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel