missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98973-100ul
This item is not returnable.
View return policy
Description
HSD17B4 Polyclonal antibody specifically detects HSD17B4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HSD17B4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| 12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, 7-alpha, D-bifunctional protein, EDH17B4, hydroxysteroid (17-beta) dehydrogenase 4, MPF-2, peroxisomal, peroxisomal multifunctional enzyme type 2,17-beta-HSD IV, peroxisomal multifunctional protein 2,17beta-estradiol dehydrogenase type IV, short chain dehydrogenase/reductase family 8C, member 1,3-alpha | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HSD17B4. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3295 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction