missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD3B7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14103
This item is not returnable.
View return policy
Description
HSD3B7 Polyclonal specifically detects HSD3B7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HSD3B7 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:500 | |
| 3 beta-hydroxy-delta 5-C27-steroid oxidoreductase, 3 beta-hydroxysteroid dehydrogenase type 7, 3 beta-hydroxysteroid dehydrogenase type VII, 3-beta-HSD VII, 7-alpha-diol 3-beta-dehydrogenase, C(27) 3-beta-HSD, C(27)-3BETA-HSD, Cholest-5-ene-3-beta, EC 1.1.1.-, EC 1.1.1.181, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7,3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase, PFIC4, SDR11E3, short chain dehydrogenase/reductase family 11E, member 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80270 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HSD3B7 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VVRMLLQREPRLGELRVFDQHLGPWLEELKTGPVRVTAIQGDVTQAHEVAAAVAGAHVVIHTAGLVDVFGRASPKTIHEVNVQG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction