missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF2 Antibody (SR2042), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22062-100ul
This item is not returnable.
View return policy
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| HSF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR2042 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
| A synthesized peptide derived from human HSF2 (Uniprot #: Q03933) | |
| 100 μg | |
| Cellular Signaling, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 3298 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction