missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF4 Antibody (2E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37499
This item is not returnable.
View return policy
Description
HSF4 Monoclonal specifically detects HSF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| HSF4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| cataract, Marner, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSTF 4 | |
| Mouse | |
| 53 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 20000000 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HSF4 | |
| Purified recombinant fragment of human HSF4 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 3299 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction