missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSF5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15857893
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15857893 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15857893 Supplier Invitrogen™ Supplier No. PA569484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%.

HSF5 may act as a transcriptional factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSF5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hsf5
Gene Accession No. Q4G112
Gene Alias Gm739; heat shock factor protein 5; Heat shock transcription factor 5; heat shock transcription factor family member 5; HSF 5; HSF5; HSTF 5; HSTF5; LOW QUALITY PROTEIN: heat shock factor protein 5; MLZ-220; Protein expressed in male leptotene and zygotene spermatocytes 220
Gene Symbols Hsf5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human HSF5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124535
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.