missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp47 Antibody (SR1796), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21700-100ul
This item is not returnable.
View return policy
Description
Hsp47 Monoclonal antibody specifically detects Hsp47 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Hsp47 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1796 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200 | |
| (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
| A synthesized peptide derived from human Hsp47 (Uniprot #: P50454) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 871 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction