missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marque: Novus Biologicals NBP1-77396SS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HSP60 Polyclonal specifically detects HSP60 in Human, Mouse, Rat, Hamster, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Spécification
| HSP60 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 30% Glycerol with 0.05% Sodium Azide | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, hamster, and primate. Predicted to react with bovine based on 100% sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 0.5 ug/ml, Simple Western 1:5000, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:200 | |
| P10809, P63038 | |
| HSPD1 | |
| A synthetic peptide made to an internal region of the human Hsp60 protein (within residues 300-360). [Swiss-Prot P10809] | |
| 0.025 mL | |
| Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 3329 | |
| Human, Mouse, Rat, Hamster, Primate | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu