missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
355.00 € - 518.00 €
Specifications
| Antigen | HSP60 |
|---|---|
| Dilution | Western Blot 1:10000-1:20000, Immunohistochemistry 1: 2000-1:5000, Immunocytochemistry/ Immunofluorescence 1:2000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627736
|
Novus Biologicals
NBP3-05536-100ul |
100 μg |
518.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612978
|
Novus Biologicals
NBP3-05536-50ul |
50 μg |
355.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP60 Polyclonal antibody specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| HSP60 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| Supplied as an aliquot of IgY preparation | |
| 3329 | |
| IgY | |
| IgY purified |
| Western Blot 1:10000-1:20000, Immunohistochemistry 1: 2000-1:5000, Immunocytochemistry/ Immunofluorescence 1:2000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Recombinant full length human HSP60 expressed in and purified from E. coli | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title