missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00 € - 498.00 €
Specifications
| Antigen | HSP90 beta |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611087
|
Novus Biologicals
NBP2-68978-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651978
|
Novus Biologicals
NBP2-68978 |
100 μg |
498.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP90 beta Polyclonal antibody specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| HSP90 beta | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| PBS (pH 7.2) and 40% Glycerol | |
| 3326 | |
| IgG | |
| Protein A purified |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EEDKDDEEKPKIEDVGSDEEDDSGKDK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title