missing translation for 'onlineSavingsMsg'
Learn More

Hsp90 Mouse, Unlabeled, Clone: 68, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610419

 View more versions of this product

Product Code. 15865449

  • ONLINE EXCLUSIVE PRICE
    336.00 €
    323.14 € / 1 set
    Save 12.86 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 90kDa heat shock protein (Hsp90), a highly conserved stress-induced protein, is abundantly expressed in most tissues under nonstress conditions and is required for eukaryotic cell viability. Hsp90 interacts with a number of signaling molecules, other heat shock proteins, and cytoskeletal proteins. However, the function of Hsp90 in these complexes remains unclear. It is thought that Hsp90 plays a common role in a number of diverse signaling systems due to its physical association with v-Src family tyrosine kinases, steroid hormone receptors, the Raf serine/threonine kinase, the basic-helix-loop-helix dioxin receptor, and the βγ subunits of G proteins.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hsp90
Monoclonal
250μg/mL
Heat Shock Protein-90
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
68
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.