missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA12B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88294-25ul
This item is not returnable.
View return policy
Description
HspA12B Polyclonal specifically detects HspA12B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HspA12B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| C20orf60, chromosome 20 open reading frame 60, dJ1009E24.2, FLJ32150, heat shock 70 kDa protein 12B, heat shock 70kD protein 12B, MGC131912 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 116835 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HSPA12B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LAVPEMGLQGLYIGSSPERSPVPSPPGSPRTQESCGIAPLTPSQSPKPEVRAPQQASFSVVVAIDFGTTSSGYAFSFASDPEAIHMMRKWEGG | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering