missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA1A Recombinant Rabbit Monoclonal Antibody (24GB3205)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557040
This item is not returnable.
View return policy
Description
HSPA1A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
Specifications
| HSPA1A | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0DMV8, P0DMW0, Q61696 | |
| HSPA1A | |
| A synthesized peptide derived from human Hsp70 (aa 400-641). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3205 | |
| Unconjugated | |
| HSPA1A | |
| 68 kDa heat shock protein; APG2; APG-2; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa protein 4; heat shock 70kD protein 1A; heat shock 70kD protein 4; heat shock 70kDa protein 1A; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; heat shock protein 1A; heat shock protein 70; heat shock protein 70-1; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; heat shock protein, 70 kDa 3; heat shock-induced protein; HEL-S-103; HEL-S-5a; HS24/P52; hsp68; hsp70; hsp70 RY; HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; Hsp70-2; Hsp70-3; Hsp70A1; HSP70I; hsp70RY; HSP72; Hspa1; HSPA1A; Hspa1b; Hspa2; HSPA4; HSPH2; HSX70; inducible heat shock protein 70; RY | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 193740, 24472, 3303 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction