missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA6 Antibody (165f), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97466-0.025MG
This item is not returnable.
View return policy
Description
HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Western Blot.
Specifications
| HspA6 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| HSPA6 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of human HspA6 Antibody (165f). | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 165f | |
| Western Blot 1:1000 | |
| heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B' | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3310 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction