missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSR3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97569AF647
This item is not returnable.
View return policy
Description
SSR3 Polyclonal specifically detects SSR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SSR3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 6747 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| signal sequence receptor, gamma (translocon-associated protein gamma), SSR gamma, SSR-gamma, translocon-associated protein gamma subunit, translocon-associated protein subunit gamma, TRAP-complex gamma subunit, TRAP-gamma, TRAPGSignal sequence receptor subunit gamma | |
| Produced in rabbits immunized with E. coli-derived Human SSR3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction