missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspB2 Antibody (4B9E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52490-0.025mg
This item is not returnable.
View return policy
Description
HspB2 Monoclonal specifically detects HspB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| HspB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| DMPK-binding protein, HspB2, MKBP | |
| Mouse | |
| 20.2 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| 4B9E2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HSPB2 | |
| Purified recombinant fragment of human HSPB2 (AA: 1-182) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 3316 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction