Learn More
Invitrogen™ HTR1A Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA595035
Product Code. 16364305
  • 520.00 € / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human hepatocellular carcinoma tumor tissue (HCCT), rat liver tissue. IHC: human endometrial adenocarcinoma tissue, human lung cancer tissue, rat lymphaden tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HTR1A
Polyclonal
Unconjugated
HTR1A
5HT1A; 5-HT1A; 5-HT-1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A
Rabbit
Affinity chromatography
RUO
24473, 3350
-20°C
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 4mg trehalose and no preservative
P08908, P19327
HTR1A
A synthetic peptide corresponding to a sequence at the C-terminus of human 5HT1A Receptor (404-422aa NKDFQNAFKKIIKCKFCRQ).
100 μg
Primary
Human, Rat, Rat