missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HuC/HuD Monoclonal Antibody (16A11)

Product Code. 11524027
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11524027 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11524027 Supplier Invitrogen™ Supplier No. A21271
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes the Elav family members HuC, HuD and Hel-N1 neuronal proteins. It does not recognize HuR, another Elav family member that is present in all proliferating cells. The antibody has been shown to specifically label neuronal cells in zebrafish, chick, canaries, and humans, and is likely to label neuronal cells in most vertebrate species. Labeling is visible early in development, at about the time that the neurons leave the mitotic cycle. Storage and reconstitution: Upon receipt, lyophilized antibodies should be stored desiccated at -20°C or lower. When properly stored, these products are stable for at least one year. To prepare stock solution, reconstitute the antibody in 0.5-1 mL PBS, pH 7.4, containing 1% BSA. Store the solution for up to two weeks at 4°C with the addition of 2mM sodium azide. For longer storage, divide solutions into single-use aliquots and freeze at -20°C, avoidig freeze/thaw cycles. For best results in immunohistochemistry, antigen retrieval is required. Use 50mM Tris, pH 8.0, for 30 minutes. A-21271 was successfully used to detect HuC/HuD in neurons differentiated from H9 ESC derived NSCs.

Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HuC/HuD
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 16A11
Conjugate Unconjugated
Formulation HEPES buffered saline with no preservative
Gene ELAVL4
Gene Accession No. P26378, Q14576
Gene Alias ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 3; ELAV like neuron-specific RNA binding protein 4; ELAV like RNA binding protein 4; elavC; ELAVL3; elavl3 protein; elavl4; ELAV-like protein 3; ELAV-like protein 4; elav-related C; elav-related D; elrc; elrd; embryonic lethal, abnormal vision-related C; embryonic lethal, abnormal vision-related D; etID309927.3; Hu antigen C; Hu antigen D; hu-antigen C; Hu-antigen D; HuC; HUCL; hud; id:ibd1248; Paraneoplastic cerebellar degeneration-associated antigen; paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; PLE21; PNEM; protein elrD; ribonucleoprotein; RNA-binding protein HuD; wu:fb77b03; wu:fc24h01; zHuC
Gene Symbols ELAVL3, ELAVL4
Host Species Mouse
Immunogen Human HuC/HuD neuronal protein.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1995, 1996, 30732, 30737, 395634
Target Species Avian, Chicken, Human, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.