missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 20S Immunoproteasome Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8194-100
This item is not returnable.
View return policy
Description
20S Immunoproteasome Monoclonal specifically detects 20S Immunoproteasome in Human samples. It is validated for Immunocytochemistry.
Specifications
| 20S Immunoproteasome | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human 20S Immunoproteasome purified from peripheral blood mononuclear cells | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 894510 | |
| Immunocytochemistry 5-25 ug/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human 20S Immunoproteasome in direct ELISA. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction