Learn More
Human 20S Proteasome Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9554-SP
This item is not returnable.
View return policy
Description
20S Proteasome Monoclonal specifically detects 20S Proteasome in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| 20S Proteasome | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human 20S Proteasome purified from human erthrocytes | |
| 25 μg | |
| Detects human 20S Proteasome in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
| Western Blot, Immunocytochemistry | |
| 863437 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |