missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ABCG2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB995-100
This item is not returnable.
View return policy
Description
ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| ABCG2/CD338 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCG2 | |
| 3T3 cells transduced with human ABCG2 | |
| 100 μg | |
| Primary | |
| Detects human ABCG2 in flow cytometry and immunocytochemistry. | |
| Human | |
| Supernatant |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 5D3 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| ABC transporter, ABC15, ABCP, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXR, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9429 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction