missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACE-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
221.00 € - 480.00 €
Specifications
| Antigen | ACE-2 |
|---|---|
| Clone | 1034734 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723177
![]() |
R&D Systems
MAB10822-SP |
25 μg |
221.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763467
![]() |
R&D Systems
MAB10822-100 |
100 μg |
480.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACE-2 Monoclonal antibody specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| ACE-2 | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
| Mouse myeloma cell line NS0-derived human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1034734 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BYF1 | |
| 59272 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title