missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACE/CD143 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9291-SP
This item is not returnable.
View return policy
Description
ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Western Blot.
Specifications
| ACE/CD143 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1636 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 171409 | |
| Western Blot 1 ug/mL | |
| P12821.1 | |
| ACE | |
| Mouse myeloma cell line NS0-derived recombinant human ACE Leu30-Leu1261 Accession # P12821.1 | |
| 25 μg | |
| Primary | |
| Detects recombinant human (rh)ACE in direct ELISAs and Western blots. Does not cross-react with rhACE-2 or rhNeprilysin. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction