missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin RIIB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3392-500
This item is not returnable.
View return policy
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot.
Specifications
| Activin RIIB | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActivinRIIB, ActR-IIB, ACTRIIB, ACVR2B, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 93 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 60408 | |
| Western Blot 1 ug/mL | |
| CAA54671 | |
| ACVR2B | |
| Mouse myeloma cell line NS0-derived recombinant human Activin RIIB Ser19-Thr134 Accession # CAA54671 | |
| 500 μg | |
| Primary | |
| Detects human Activin RIIB in direct ELISAs and Western blots. Shows approximately 5% cross-reactivity with recombinant human Activin RIIA. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction