Learn More
Abnova™ Human ADCYAP1 Partial ORF (NP_001108.1, 95 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000116-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes adenylate cyclase activating polypeptide 1. Mediated by adenylate cyclase activating polypeptide 1 receptors, this polypeptide stimulates adenylate cyclase and subsequently increases the cAMP level in target cells. Adenylate cyclase activating polypeptide 1 is not only a hypophysiotropic hormone, but also functions as a neurotransmitter and neuromodulator. In addition, it plays a role in paracrine and autocrine regulation of certain types of cells. This gene encodes three different mature peptides, including two isotypes, a shorter form and a longer form. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: VLDQLSAGKHLQSLVARGVGGSLGGGAGDDAEPLSKRHSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNKGRRSpecifications
NP_001108.1 | |
Liquid | |
116 | |
ADCYAP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126852/PACAP | |
ADCYAP1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.32kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VLDQLSAGKHLQSLVARGVGGSLGGGAGDDAEPLSKRHSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNKGRR | |
RUO | |
ADCYAP1 | |
Wheat Germ (in vitro) | |
GST |