missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adenosine A1R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11135-100
This item is not returnable.
View return policy
Description
Adenosine A1R Monoclonal antibody specifically detects Adenosine A1R in Human samples. It is validated for Immunohistochemistry
Specifications
| Adenosine A1R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Adenosine A1 R, adenosine A1 receptor, ADORA 1, ADORA1, RDC7, RDC7adenosine receptor A1 | |
| Human Adora-1 synthetic peptide corresponding to the 2nd. extra-cellular loop.,, Accession # P30542 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1051024 | |
| Immunohistochemistry 3-25 ug/mL | |
| P30542 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 134 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction