missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 15.63 to 1000 pg/mL |
| Assay Sensitivity | 9.38 pg/mL |
| Product Line | Novus |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Serum, Plasma, Biological Fluids |
| Specificity | This kit recognizes Human ALCAM in samples. No significant cross-reactivity or interference between Human ALCAM and analogues was observed. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 214 |
| Gene Symbol | ALCAM |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?