missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3701-SP
This item is not returnable.
View return policy
Description
ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications
| ALK-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
| Mouse myeloma cell line NS0-derived recombinant human ALK-1, Asp22-Gln118, Accession # P37023 | |
| 25 μg | |
| 94 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Blocking Assay | |
| 117702 | |
| Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL | |
| P37023 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human ALK-1 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction