missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Alkaline Phosphatase/ALPL APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems FAB1448A-025
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Flow Cytometry.
Specifications
| Alkaline Phosphatase/ALPL | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| ALPL | |
| Human liver, bone and kidney-derived Alkaline Phosphatase/ALPL | |
| 25 Tests | |
| Primary | |
| Detects liver, bone and kidney Alkaline Phosphatase/ALPL from human tissue (2). | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| B4-78 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 249 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction