missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-1 Adrenergic R/ADRA1A Antibody, R&D Systems™
Description
alpha-1A Adrenergic R/ADRA1A Monoclonal specifically detects alpha-1A Adrenergic R/ADRA1A in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
| Antigen | alpha-1A Adrenergic R/ADRA1A |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 518508 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene Accession No. | P35348 |
| Gene Alias | a-1A AdrenergicR, ADRA1A, ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, alpha1A AdrenergicR, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?