missing translation for 'onlineSavingsMsg'
Learn More

Human alpha-Smooth Muscle Actin Alexa Fluor 594-conjugated Antibody, R&D Systems™

Product Code. p-200039023
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18154321 100 Tests 100 tests
18757509 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18154321 Supplier R&D Systems Supplier No. IC1420T100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Flow Cytometry
Classification Monoclonal
Clone 1A4
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin
Gene Symbols ACTA2
Host Species Mouse
Immunogen alpha-Smooth Muscle Actin N-terminal Synthetic Decapeptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59
Test Specificity Detects human alpha-Smooth Muscle Actin.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.