missing translation for 'onlineSavingsMsg'
Learn More

Human ASCL2/Mash2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18708527
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18708527 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18708527 Supplier R&D Systems Supplier No. AF6539SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ASCL2/Mash2 Polyclonal specifically detects ASCL2/Mash2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL2/Mash2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99929
Gene Alias achaete-scute complex (Drosophila) homolog-like 2, achaete-scute complex homolog 2 (Drosophila), achaete-scute complex-like 2 (Drosophila), ASH2, ASH-2, BHLHA45, bHLHa45achaete-scute homolog 2, Class A basic helix-loop-helix protein 45, HASH2, HASH2achaete-scute complex-like 2, mammalian achaete/scute homologue 2, MASH2
Gene Symbols ASCL2
Host Species Sheep
Immunogen E. coli-derived recombinant human ASCL2/Mash2 Met1-Ala49 Accession # Q99929
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 430
Test Specificity Detects human ASCL2/Mash2 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.