missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
203.00 € - 479.00 €
Specifications
| Antigen | B7-H4 |
|---|---|
| Clone | 2318A |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732287
![]() |
R&D Systems
MAB65764-SP |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743817
![]() |
R&D Systems
MAB65764-100 |
100 μg |
479.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H4 Monoclonal specifically detects B7-H4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| B7-H4 | |
| Western Blot 1 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
| Mouse myeloma cell line, NS0-derived human B7-H4, Phe29-Ser258, Accession # Q7Z7D3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human B7-H4 in direct ELISAs. |
| 2318A | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Human | |
| Q7Z7D3 | |
| 79679.0 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title