missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bad Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6405-SP
This item is not returnable.
View return policy
Description
Bad Monoclonal specifically detects Bad in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
Specifications
| Bad | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 5-25 ug/mL, Knockout Validated | |
| Q92934 | |
| BAD | |
| E. coli-derived recombinant human Bad Met1-Gln168 Accession # Q92934 | |
| 25 μg | |
| Primary | |
| Detects human Bad in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Bad (aa 1-130), rmBcl-2 alpha , recombinant human (rh) Bcl-2, or rhBCL2L12 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
| 612312 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 572 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction