missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BCMA/TNFRSF17 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB193-SP
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| BCMA/TNFRSF17 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q02223 | |
| TNFRSF17 | |
| Mouse myeloma cell line NS0-derived recombinant human BCMA Met1-Ala54 Accession # Q02223 | |
| 25 μg | |
| Primary | |
| Detects human BCMA in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with recombinant mouse (rm) BCMA or rmTACI. | |
| Human | |
| Purified |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 335004 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 608 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction