missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB648-500
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, ELISA.
Specifications
| BDNF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 627 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 37141 | |
| Western Blot 1 ug/mL, ELISA | |
| P23560 | |
| BDNF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF Arg128-Arg247 Accession # P23560 | |
| 500 μg | |
| Primary | |
| Detects human BDNF in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) NT-3 or rhNT-4. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction