missing translation for 'onlineSavingsMsg'
Learn More

Human Betacellulin/BTC Antibody, R&D Systems™

Product Code. 18783407
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18783407 25 μg 25µg
18720487 100 μg 100µg
18770757 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18783407 Supplier R&D Systems Supplier No. MAB261SP
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    197.40 € / 25µg
    Save 25.64 €
    11% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Human samples. It is validated for ELISA Capture (Matched Antibody Pair)∣ELISA Development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Betacellulin/BTC
Applications ELISA Capture (Matched Antibody Pair), ELISA Development
Classification Monoclonal
Clone 34362
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35070
Gene Alias betacellulin, BTC, probetacellulin
Gene Symbols BTC
Host Species Mouse
Immunogen E. coli-derived recombinant human Betacellulin Asp32-Tyr111 Accession # P35070
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 685
Test Specificity Detects human Betacellulin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) EGF, rhHB-EGF, rhTGF-alpha , rhHRG-alpha , or rhAR under non-reducing conditions.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.