missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BPI Antibody, R&D Systems™

Description
BPI Monoclonal specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry.
Specifications
Specifications
| Antigen | BPI |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 971526 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P17213 |
| Gene Alias | bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, BPIFD1, CAP 57, rBPI |
| Host Species | Mouse |
| Show More |
For Research Use Only (RUO)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?