missing translation for 'onlineSavingsMsg'
Learn More

Human Bradykinin RB2/BDKRB2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

203.00 € - 435.00 €

Specifications

Antigen Bradykinin RB2/BDKRB2
Clone 471916
Dilution Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18704417
GREENER_CHOICE
View Documents
R&D Systems
MAB47251-SP
25 μg
203.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18773207
GREENER_CHOICE
View Documents
R&D Systems
MAB47251-100
100 μg
435.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications

Specifications

Bradykinin RB2/BDKRB2
Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready
Monoclonal
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088
NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession No. P30411
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Bradykinin RB2/BDKRB2 in direct ELISAs.
471916
Flow Cytometry, Immunohistochemistry, CyTOF
Unconjugated
Human
P30411
624.0
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.