missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTNL8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB93591-SP
This item is not returnable.
View return policy
Description
BTNL8 Monoclonal antibody specifically detects BTNL8 in Human samples. It is validated for Immunohistochemistry
Specifications
| BTNL8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347 | |
| Human embryonic kidney cell HEK293-derived human BTNL8 protein, Gln18-Lys238, Accession # Q6UX41-1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1047350 | |
| Immunohistochemistry 5-25 ug/mL | |
| Q6UX41-1 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 79908 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction